BioReady Gold Nanospheres fuctionalized with Streptavidin. B, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AUIR40-10M AUIR40-1M AUIR40-200U AUIR40-5M AUIR40-10M AUIR40-1M AUIR40-200U AUIR40-5M
103819-900EA 3215 USD
103819-900 103819-902 103819-904 103819-906 77680-074 77680-076 77680-078 77680-080
BioReady Gold Nanospheres fuctionalized with Streptavidin. B, nanoComposix
Beads Microcarriers
40 nm gold nanoparticles functionalized with streptavidin
  • Streptavidin tetramer is covalently bound to the particle surface via an amide bond. Streptavidin gold conjugates can be used for convenient conjugation of any biotinylated antibody
Streptavidin tetramer is covalently bound to the particle surface via an amide bond. Streptavidin gold conjugates can be used for convenient conjugation of any biotinylated antibody, protein or oligonucleotide.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, and UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Información de suministro: Ships at ambient temperatures
Precaución: Do not freeze. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.