BioReady Gold Nanospheres for Conjugation, nanoComposix
Supplier: NANOCOMPOSIX, INC.
AUCR40-100M AUCR40-30M AUCR40-5M AUCR40-100M AUCR40-30M AUCR40-5M
103819-864EA 1360 USD
103819-864 103819-866 103819-868 77680-030 77680-032 77680-034
BioReady Gold Nanospheres for Conjugation, nanoComposix
Beads Microcarriers
40 nm gold nanoparticles for highly reproducible gold conjuates for assays and diagnostics.
  • Achieve reliable gold conjugates for reproducible results in diagnostics and lateral flow assays
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
BioReady materials are engineered for reliable conjugation to antibodies, proteins, peptides, and other biomolecules. Ideal for lateral flow and other detection and diagnostics.
Each batch is extensively characterized to meet highest quality standards,More Product Information