Ritonavir ≥98% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
Synonyms:
A-84538, Thiazol-5-ylmethyl ((2S,3S,5S)-3-hydroxy-5-((S)-2-(3-((2-isopropylthiazol-4-yl)methyl)-3-methylureido)-3-methylbutanamido)-1,6-diphenylhexan-2-yl)carbamate, ABT-538, Abbott 84538
R0116-1G
R0116-200MG
TCR0116-1GEA
186.2
USD
TCR0116-1G
TCR0116-200MG
Ritonavir ≥98% (by HPLC, total nitrogen)
Ritonavir
Formula:
C₃₇H₄₈N₆O₅S₂ |
MDL Number:
MFCD00927142 CAS Number: 155213-67-5 |
|
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity(HPLC) | min. 98.0 area% |
Purity(with Total Nitrogen) | min. 98.0 % |
Melting point | 122.0 to 127.0 deg-C |
Specific rotation [a]20/D | +7.0 to +10.0 deg(C=2,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...