Nalgene® Biotainer Bottles, Square, PETG, Clean, Platinum Certified, Thermo Scientific
Supplier: Thermo Fisher Scientific
CE-N3005-42 CE-N3025-42 CE-N3110-42 CE-N3230-42
76412-168CS 2854.15 USD
76412-168 76412-170 76412-172 76412-174
Nalgene® Biotainer Bottles, Square, PETG, Clean, Platinum Certified, Thermo Scientific
Bottles Plastic Bottles
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Durable, translucent PETG construction ensures the integrity of samples
  • Closures feature a silicone liner for leakproof assurance
  • Customization options are available for fluid transfer applications
  • Extensive validation documentation support
  • Suitable for use in storage conditions of –100 to 100 °C
Bottles are washed with USP water for injection and handled post-washing in Class 5 cleanrooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than ¹/₃ the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10⁻⁶ inside the container/closure, including two heat-sealed sterile barriers to protect the product prior to use.