Tek-Select® Autoslip Coverslip Preferred Coverglasses, IMEB
Supplier: IMEB INC MS
AS-2450 AS-2455 AS-2440
103783-104CS 98.94 USD
103783-104 103783-106 103783-108
Tek-Select® Autoslip Coverslip Preferred Coverglasses, IMEB
Cover Glasses
Tek-Select® Autoslips coverslips are specifically designed for use on instruments that automatically apply cover glass.
  • Uniform No. 1 thickness
  • Multiple sizes available
Tek-Select® Autoslips coverslips also perform well for manual coverslipping in humid environments.