Formalin 10% with zinc, Tek-Select®, buffered
Supplier: IMEB INC MS
Synonyms:
Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
Tek-Select® 10% Buffered Formalin with Zinc is prepared using only high purity chemicals and is ready for application and can be easily used in office-based sites as well as clinics.
- Ready for application
- Prepared using only high purity chemicals
- 10% Formalin solution with 1% Zinc Sulfate and Acetate buffer added in deionized water
- Acetate buffer stabilizes the pH of solution and prevents the possible formation of formalin pigment
It is recommended as an all-purpose tissue fixative for demonstration of crisp nuclear detail, superior cellular morphology, enhanced Hematoxylin and Eosin staining, special staining and immunohistochemical studies.
It also provides excellent results for lymph nodes and gastric biopsies.
Tek-Select® 10% Buffered Formalin with Zinc presents minimal safety hazards and is non-corrosive.
Formula:
CH₂O MW: 30.03 g/mol |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...