tert-Amyl methyl ether ≥99.0% (by GC) produced by INEOS Oligomers

Danger

Synonyms: TAME, METHOXYPENTANE™, Methyl 2-methyl-2-butyl ether, 1,1-Dimethylpropyl methyl ether, Methyl tert-pentyl ether

742619-1L 742619-5L
102325-288EA 140 USD
102325-288 102325-290
tert-Amyl methyl ether ≥99.0% (by GC) produced by INEOS Oligomers
tert-Amyl methyl ether

METHOXYPENTANE is a trademark of INEOS Oligomers.

Formula: C₆H₁₄O
MW: 102.17 g/mol
Boiling Pt: 85…86 °C (lit.)
Density: 0.76…0.78 g/mL (25 °C) (lit.)
Flash Pt: –7 °C
MDL Number: MFCD00010186
CAS Number: 994-05-8
EINECS: 213-611-4
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR