Micro Graham Condenser, Chemglass
Supplier: Chemglass
CG-MR-50C2
76333-428EA 368.29 USD
76333-428
Micro Graham Condenser, Chemglass
Condensers
Micro Graham condenser with 101 mm OAL, GL-14 top thread, 10/18 lower inner joint, and GL-14 thread inlet/outlet.
Información de suministro: Supplied complete with caps with apertures and hose connections.