Corepoint Scientific™ General Purpose Flammable Storage Refrigerators, Horizon Scientific
Supplier: Horizon Scientific
FR301WWW/0GP FRF302WWW/0GP
76402-986EA 7781.99 USD
76402-986 76402-988
Corepoint Scientific™ General Purpose Flammable Storage Refrigerators, Horizon Scientific
Refrigerators
These flammable storage units are designed to store and protect flammable substances in laboratory settings.
  • Mechanical thermostat
  • Interior shelving
  • Keyed door lock
  • Probe access
  • Meet NFPA guidelines
These refrigerators are designed to store and protect volatile substances in laboratory settings. These units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
Información para pedidos: All units come with a one year parts and labor warranty.