Buffers, Precision Reference Standards, RICCA Chemical Company
Supplier: Ricca Chemical
R1602000-500G
R1502000-500G
R1552000-500G
1513-16
1532-16
1572-16
R1493500-500G
RC160216CS
1152
USD
RC160216
RC1502-16
RC1552-16
RC1513-16
RC1532-16
RC1572-16
75832-314
Buffers, Precision Reference Standards, RICCA Chemical Company
Buffers
pH Calibration Buffers
This precision line of pH Buffers offers tolerances of ±0.005 pH unit or better.
Certificaciones: Compared against and certified traceable to NIST Standard Reference Materials.