Broadband IR-CO₂, Glass Lens LaserShields, NoIR Laser Company
Supplier: NOIR MEDICAL
FG137 FG146
76332-480EA 312.84 USD
76332-480 76332-482
Broadband IR-CO₂, Glass Lens LaserShields, NoIR Laser Company
Safety Glasses
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certificaciones: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Información para pedidos: Hard-shell case, cleaning cloth, and strap included.