KIMBLE® Extended Body Vacuum Filtration Adapters with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
178120-0014 178120-0024 178120-0124
KT178120-0014EA 141.2 USD
KT178120-0014 KT178120-0024 KT178120-0124
KIMBLE® Extended Body Vacuum Filtration Adapters with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Vacuum filtration adapters with hose connection is used for reduced pressure filtration using plain stem buchner funnels.
  • Designed with a flange to accommodate a pluro stopper
  • Includes a lower drip tip
Manufactured from 33 expansion, low extractable borosilicate glass.
Certificaciones: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Información para pedidos: Pluro stopper sold separately.