DURAN® GLS 80® High Temperature Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
1152921 1165888 1167307
76263-772PK 118.29 USD
76263-772 76263-780 76264-164 76264-166
DURAN® GLS 80® High Temperature Screw Caps, DWK Life Sciences
Closures Bottle Closures
The material used is a special compound based on polyarylsulphone. Consequently the chemical, thermal and mechanical properties of the material are noticeably improved and matched to laboratory requirements.
  • PSU material
  • Autoclavable
  • With cap liner
The DURAN® GLS 80® bottle can be opened or closed with only a three-quarter turn. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly sealed (cap liner: Platinium-cured silicone). A matching PTFE pouring ring is also available, permitting clean, drip-free use.