Memphis Welding Leather Welding Sleeves with Adjustable Neck Straps, MCR Safety
Supplier: MCR Safety
38423MW 38425MW
76341-012PR 90.7 USD
76341-012 76341-082
Memphis Welding Leather Welding Sleeves with Adjustable Neck Straps, MCR Safety
Workwear
Heavy select gray side-split leather provides outstanding protection from heat, sparks, slag, and cuts.
  • Select gray split cow leather
  • Double lock stitched, sewn with Kevlar®
  • Sleeve with adjustable neck strap, 23 or 25 Inch length
Sleeves are ideally suited for use in welding, construction, Heliarc, iron work, metal fabrication and Mig/Tig applications.