BBL™ Trichophyton Agar, BD Life Sciences
Supplier: BD
296246 296245
90004-674PK 96.02 USD
90004-674 90004-680 90004-918 90004-678
BBL™ Trichophyton Agar, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Trichophyton Agars are differential media used in the presumptive identification of Trichophyton species based on nutritional requirements.
  • Supplied as prepared slants in C tubes