Thallium(III) acetate

Danger

Synonyms: Thallic acetate

151165-25G 151165-5G
220036-973EA 1719.38 USD
220036-973 220036-974
Thallium(III) acetate
Thallium(III) acetate
Formula: (C₂H₄O₂)₃Tl
MW: 381.52 g/mol
MDL Number: MFCD00008698
CAS Number: 2570-63-0
ADR: 6.1,II

Order Now

Specification Test Results

Appearance (Color) Conforms to Requirements
White to Off-White or Light Peach
Appearance (Form) Powder or Crystals
Infrared Spectrum Conforms to Structure
Assay % Tl (ICP) 51.0 - 56.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR