Anti-E. protein Mouse Monoclonal Antibody [clone: E60]

Supplier: Bio X Cell
BE0325-1MG BE0325-25MG BE0325-5MG BE0325-100MG BE0325-50MG
103778-468EA 3920 USD
103778-468 103778-470 103778-472 103778-474 103778-476 76554-338 76554-584 76554-808 76555-464 76556-406
Anti-E. protein Mouse Monoclonal Antibody [clone: E60]
Antibodies
Type: Primary
Antigen: E. protein
Clonality: Monoclonal
Clone: E60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: West Nile, dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR