Anti-Ganglioside GD2 Mouse Monoclonal Antibody [clone: 14G2a]

Supplier: Bio X Cell
BE0318-50MG BE0318-1MG BE0318-25MG BE0318-100MG BE0318-5MG
103778-398EA 3920 USD
103778-398 103778-400 103778-402 103778-404 103778-406 76555-050 76555-320 76555-440 76555-510 76557-112
Anti-Ganglioside GD2 Mouse Monoclonal Antibody [clone: 14G2a]
Antibodies
Type: Primary
Antigen: Ganglioside GD2
Clonality: Monoclonal
Clone: 14G2a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR