in vivo-jetRNA® Transfection Reagents, Polyplus-transfection
76358-644EA 713.6 USD
76358-644 76358-646
in vivo-jetRNA® Transfection Reagents, Polyplus-transfection
Transfection Reagents
in vivo-jetRNA® is an injectable in vivo mRNA transfection reagent, allowing an adaptable, safe, time-saving and tailored animal injection.
  • Adaptable: Suited for any injection route to target any organ
  • Safe: Maintain healthy animals and organs
  • Time-saving: Ready-to-use reagent with an easy protocol
  • Tailored: Customized protocols from in vivo delivery experts
in vivo-jetRNA® is a transfection reagent specifically developed to deliver mRNA in vivo. This can be used to target multiple organs, by using systemic or local injection routes, in various animal models. mRNA delivery using in vivo-jetRNA® is user-friendly, with a simple two-step protocol and can be used for vaccination purposes, anti-cancer studies, genome editing using CRISPR/Cas9 method or protein replacement.
Certificaciones: Polyplus-transfection® is ISO 9001 certified.