Iona Lite Pants, Portwest
Supplier: Portwest
S481NARM S481NARS S481NARXL S481NARXXL
76104-566EA 33.81 USD
76104-566 76104-568 76104-570 76104-572
Iona Lite Pants, Portwest
Workwear
Durable and waterproof pants with sealed seams ensure a complete barrier against wind and rain.
  • Reflective tape for visibility
  • Two side pass-through pockets
  • Fully elasticated waist
  • Abrasion resistant polyester
Snap adjustable hem with generous fit for wearer comfort.
Certificaciones: EN 343 Class 3:1 (WP 15.000 MVP 960 g/m²/24 hr).
Order Now
SPECIFICATIONS
Model S481
Garment application Mechanical protection,Industrial,Outdoor
Environmentally Preferable
Risk area HiVis,Rain (waterproof)
Gender Unisex
Sleeve or leg length Long
Fabric 100% polyester, 300D Oxford weave with stain resistant finish, PU coated
Weight 190 g