IGEPAL® CA-630

Danger

Synonyms: Octylphenyl-polyethylene glycol

56741-250ML-F 56741-50ML-F
220079-284EA 210.77 USD
220079-284 220079-285
IGEPAL® CA-630
IGEPAL® CA-630
Formula: (C₂H₄O)nC₁₄H₂₂O
Flash Pt: 251 °C
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82500c8eb1edf6c10e7d2f228334d6e01bf67bf8&vid=3666cd53b279d94e46a5c4497718c6ecdf658479&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR