IGEPAL® CA-630, viscous liquid

Danger

Synonyms: Octylphenyl-polyethylene glycol

I3021-50ML I3021-100ML I3021-500ML
220192-728EA 57.78 USD
220192-728 220192-726 220192-727
IGEPAL® CA-630, viscous liquid
IGEPAL® CA-630
Formula: (C₂H₄O)nC₁₄H₂₂O
Flash Pt: 251 °C
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR