IGEPAL® CA-630 for molecular biology

Danger

Synonyms: Octylphenyl-polyethylene glycol

I8896-50ML I8896-100ML
220193-351EA 78.17 USD
220193-351 220193-350
IGEPAL® CA-630 for molecular biology
IGEPAL® CA-630
Formula: (C₂H₄O)nC₁₄H₂₂O
Flash Pt: 251 °C
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR