Paraformaldehyde 1% in 0.1 M phosphate buffer pH 7.4 fixative reagent

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

15952-10 15952-15
102091-938PK 81.27 USD
102091-938 102091-942
Paraformaldehyde 1% in 0.1 M phosphate buffer pH 7.4 fixative reagent
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
Merck Index: 13,04259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f240682c03df85e943acbb1a983fe71750db44cd&vid=96c37abe2cf4c6160dda3947fb05d9d23c32fa74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR