Trimethylamine - (CH₃)₃NH+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010950-100N RV010950-250N RV010950-500N
RCRV010950100NEA 413 USD
RCRV010950100N RCRV010950250N RCRV010950500N
Trimethylamine - (CH₃)₃NH+ Standard, for Ion Chromatography, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Trimethylammonium Standards
  • NIST traceable
Certificaciones: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Precaución: 1000 ppm =1 g/L= 1000 µg/ml