Tellurium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010320-100N
RV010320-250N
RV010320-500N
RV010491-100N
RCRV010320-50NEA
81.28
USD
RCRV010320-50N
RCRV010320100N
RCRV010320250N
RCRV010320500N
RCRV010421100N
RCRV010421250N
RCRV010421500N
RCRV010491-50N
RCRV010491100N
RCRV010491250N
RCRV010491500N
Tellurium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Tellurium Standards
- NIST traceable
Certificaciones: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Precaución: 1000 ppm =1 g/L= 1000 µg/ml