Tellurium Standards, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010055-100N
RV010055-500N
RV010085-100N
RV010085-500N
RCRV010055100NEA
80
USD
RCRV010055100N
RCRV010055500N
RCRV010085100N
RCRV010085500N
Tellurium Standards, for AAS, VeriSpec®, Ricca Chemical Company
Standards
Cationic Standards
Tellurium Standards
- NIST traceable
Certificaciones: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Precaución: 1000 ppm =1 g/L= 1000 µg/ml