Cadmium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010372-250N RV010372-500N RV010372-100N
RCRV010372250NEA 324.03 USD
RCRV010372250N RCRV010372500N RCRV010372100N
Cadmium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Cadmium Standards
  • NIST traceable
Certificaciones: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Precaución: 1000 ppm =1 g/L= 1000 µg/ml