Baths, Stainless Steel, JULABO
Supplier: JULABO
9903413 9903417 9903419 9903427 9903433 9903439
10015-832EA 1197.46 USD
10015-832 10015-834 10015-836 10015-838 10015-840 10015-842
Baths, Stainless Steel, JULABO
Baths Water Baths, Unstirred
Stainless steel bath tanks for use with heating immersion circulator up to +150 °C.
  • Insulated stainless steel tanks
  • Integrated handles in the rim
  • Suitable for use with CORIO C/CD heating immersion circulators
These tanks are used for sample incubation in conjunction with heating immersion circulators.
Información para pedidos: Tank volumes available in 5, 13, 17, 19, 27, 33 and 39 L sizes.