KleenGuard™ Nemesis™ Rx Readers Prescription Glasses, Kimberly-Clark Professional
Supplier: Ansell Healthcare
28618 28621 28624 28627 28630
76193-572CS 88.54 USD
76193-572 76193-574 76193-576 76193-578 76193-580
KleenGuard™ Nemesis™ Rx Readers Prescription Glasses, Kimberly-Clark Professional
Safety Glasses
Popular KleenGuard™ Nemesis™ safety glasses are equipped with varied magnification strengths to suit those who need vision assistance.
  • Black frame with clear lenses, ideal for indoor use
  • Stylish nylon frames and self-adjusting rubber nose pad for comfort and fit
  • Sporty, flexible, lightweight design
  • FLEX-DRY™ nosepiece designed to channel sweat away from the eyes and prevent slippage
  • Includes a convenient neck cord, glasses are ready to protect at a moment’s notice
The Nemesis™ Rx readers re designed for long-term comfort, thanks to flexible and lightweight design, soft touch temples, and a nosepiece with integrated ridges to channel away sweat. These safety glasses have polycarbonate lenses that provide 99.9% UVA/UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss. The wraparound frame has a base curve of eight that extends the range of peripheral protection. The corrective lenses are hard coated for scratch resistance and longer life.
The safety eyewear is manufactured to comply with the ANSI/ISEA Z87.1+ standard for personal eye protection. Both men and women will find these stylish glasses to be comfortable and functional for construction, manufacturing, and a host of other industries.
Certificaciones: Complies with the ANSI / ISEA Z87.1+ standard for personal eye protection and certified to meet the requirements of CSA Z94.3.
Información de suministro: Each pair of KleenGuard Nemesis safety glasses come with a neck cord for securing them, so that the glasses are ready to protect at a moment’s notice.