KleenGuard™ G80 Neoprene/Latex Chemical Resistant Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
38741 38742 38743 38744
76300-884CS 384.81 USD
76300-884 76300-886 76300-888 76300-890
KleenGuard™ G80 Neoprene/Latex Chemical Resistant Gloves, Kimberly-Clark
Gloves
KleenGuard™ G80 neoprene/latex chemical resistant gloves provide the ultimate in chemical protection, comfort, and durability.
  • Neoprene over natural rubber latex construction
  • Cotton flock lining for comfort and perspiration absorption
  • High resistance to acids, oils, solvents, and alkali
KleenGuard™ G80 neoprene/latex chemical resistant gloves feature a natural rubber latex construction, covered in neoprene, which gives them high resistance to acids, alkalis, oils and solvents. These chemical gloves are 27.5 mil thick with a generous 12" length. They have a cotton flock linking, which adds comfort and absorbs perspiration for easy donning and removal.
These gloves contain natural rubber latex, they are not suitable for those with type 1 latex allergies. They are a smart choice for cleaning and degreasing, solvent wiping for parts and machinery, handling acids, bases, alcohols, alkalis, phenols and salts, and general chemical processing.
Certificaciones: Glove meets the EN 374-AKL standard and have a high level of dexterity, per EN 420:2003 standard.
Envase: Gloves are packaged in 12 pairs in a bag with 5 bags in a case.