o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1,2-Dimethylbenzene

95662-1L 95662-2.5L 95662-250ML
BJ95662-1LEA 231.77 USD
BJ95662-1L BJ95662-2.5L BJ95662-250ML
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
o-Xylene

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1815558

Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 7.6 %(V)
Melting Pt: -25 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.504 - 1.506
Non-volatile matter Max. 0.002 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
m-, p-Xylene (GC) Max. 0.6 %
Toluene (GC) Max. 0.4 %
Explosion Limit 7 %
Vapor Density 3.7 (vs air)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR