n-Pentane ≥99% (by GC), Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Pentane

60489-1L 60489-25L 60489-5L 60489-6X1L
BJ60489-1LEA 124.29 USD
BJ60489-1L BJ60489-25L BJ60489-5L BJ60489-6X1L
n-Pentane ≥99% (by GC), Riedel-de Haën™
n-Pentane

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36…37 °C
Melting Pt: –131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <–35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 500 °F
Assay (GC) Min. 99 %
Vapor Density 2.48 (vs air)
Refractive Index n20/D 1.358 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR