Glycerine solution_SC 86-89% Titrimetric, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Synonyms:
1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
49782-1L
49782-5L
BJ49782-1LEA
254.14
USD
BJ49782-1L
BJ49782-5L
Glycerine solution_SC 86-89% Titrimetric, Puriss. p.a., Riedel-de Haën™
Glycerine
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Beilstein Number: 635685
RTECS Number: 200-289-5
UNSPSC Code: MA8050000
Formula:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Boiling Pt: 290 Melting Pt: 18.6 Density: 1.26 g/cm³ Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 Merck Index: 13,04497 |
|
Specification Test Results
Assay | 85.5 - 89.5 % |
Refractive index (n 20/D) | 1.453 - 1.457 |
Sulfated ash | Max. 0.01 % |
Water (Karl Fischer) | 11 - 14 % |
Free acid (as CH₃COOH) | Max. 0.002 % |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 1 ppm |
Cobalt (Co) | Max. 1 ppm |
Chromium (Cr) | Max. 1 ppm |
Copper (Cu) | Max. 1 ppm |
Iron (Fe) | Max. 1 ppm |
Potassium (K) | Max. 20 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 1 ppm |
Sodium (Na) | Max. 20 ppm |
Nickel (Ni) | Max. 1 ppm |
Lead (Pb) | Max. 1 ppm |
Zinc (Zn) | Max. 1 ppm |
Ammonium (NH₄) | Max. 5 ppm |
Chloride (Cl) | Max. 2 ppm |
Sulfate (SO₄) | Max. 5 ppm |
Reaction against H₂SO₄ | complying |
Aldehydes (as HCHO) | Max. 0.001 % |
Fatty acid ester | Max. 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...