2,2,4-Trimethylpentane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

32291-1L 32291-2.5L 32291-4X2.5L
BJ32291-1LEA 130.18 USD
BJ32291-1L BJ32291-2.5L BJ32291-4X2.5L
2,2,4-Trimethylpentane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
2,2,4-Trimethylpentane

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1696876

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99 °C (1.013 hPa)
Melting Pt: -107 °C
Density: 0.69 g/cm³ (20 °C)
Flash Pt: -12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Boiling range 98 - 100 °C
Autoignition Temperature 745 °F
Destillation range 98-100 °C Min. 95 %
Density (D 20/4) 0.692 - 0.693
Refractive index (n 20/D) 1.3915 - 1.3925
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.0005 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
S-compounds (as S) Max. 0.005 %
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR