Petroleum spirit, 40…60 °C, Laboratory Reagent
Supplier: Honeywell Research Chemicals
|
Danger
|
15967-4X5L
15967-5L
BJ15967-4X5LCS
833.66
USD
BJ15967-4X5L
BJ15967-5L
Petroleum spirit, 40…60 °C, Laboratory Reagent
Petroleum spirit
- ≥95% for general laboratory use.
UNSPSC Code: 12191502
Boiling Pt:
40…60 °C (1.013 hPa) Density: 0.65 g/cm³ (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 265-151-9 UN: 3295 ADR: 3,II |
Specification Test Results
Boiling range | 40 - 60 °C |
Water (Karl Fischer) | Max. 0.02 % |
Non-volatile matter | Max. 0.002 % |
Aromatic substances (as C₆H₆) | Max. 0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...