i7™ High Figh Fidelity DNA Polymerase 2X Master Mix, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
i7™ high-fidelity DNA polymerase 2X master mix is a ready to use premix which contains high-fidelity DNA Polymerase, dNTPs, MgCl₂, PCR enhancers and stabilizers with optimized proprietary reaction buffer. i7™ high-fidelity DNA polymerase 2X master mix has the high-fidelity, sensitivity and processivity with an error rate ~2.8×102 fold lower than Taq DNA polymerase, and significantly lower than the error rates of other proofreading enzymes.
- Useful for long and difficult DNA amplification, high fidelity PCR, and cloning
- Supplied as convenient master mix
- Efficient for amplifying high GC template DNA with magic enhancer
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- 5X magic enhancer
i7™ High-fidelity DNA polymerase 2X master mix is ready to use premix which contains i7™ high-fidelity DNA polymerase, dNTPs, MgCl₂, PCR enhancers and stabilizers with optimized proprietary reaction buffer. i7™ high-fidelity DNA polymerase is a genetically engineered, heat stable DNA polymerase which has 5' - 3' polymerase and 3' - 5' exonuclease (proof reading) activities. This enzyme has high-fidelity, sensitivity and processivity with an error rate ~2.8×102-fold lower than Taq DNA polymerase, and significantly lower than other proof reading enzymes in the marketplace.
Información de suministro: This 2X master mix product is supplied with 5X magic enhancer that enables efficient amplification of GC rich templates up to 84%. All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.