Taq DNA Polymerase
Supplier: INTACT GENOMICS, INC MS
3245 3245D
103753-144EA 518.08 USD
103753-144 103753-146
Taq DNA Polymerase
Enzymes
High quality Taq DNA polymerase useful for routine PCR applications.
  • Useful for routine PCR, primer extension, colony PCR and more
  • Formulated for amplifying long target DNA
  • Efficient for amplifying high GC template DNA with magic enhancer
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product.
  • Taq DNA polymerase
  • 10X PCR buffer with Mg²⁺
  • 5X Magic enhancer
Taq DNA polymerase is a thermostable DNA polymerase that possesses a 5' - 3' polymerase activity and a 5' flap endonuclease activity.
Información de suministro: This product is supplied with 10X PCR reaction buffer, containing MgCl₂, which produces a final Mg²⁺ concentration of 1.5 mM. Ideal for primary extension reaction with DNA fragments having dA overhang on 3' ends. All our products are shipped with dry ice. 103753-146 includes dNTP.