Condensers, Graham, Chemglass
Supplier: Chemglass
CG-1214-01 CG-1214-03 CG-1214-05 CG-1214-07
80067-312EA 369.86 USD
80067-312 80067-316 80067-320 80067-324
Condensers, Graham, Chemglass
Condensers
Condensers feature a stop standard taper.
  • Condenser has a top standard taper outer and lower inner drip tip joint.
Hose connections have an O.D. of 10 mm at the middle serration.