Anti-IgG Rabbit Polyclonal Antibody (Biotin) PD-L1 Fc

Supplier: Peprotech
500-P321BT-25UG 500-P321BT-50UG
76304-066EA 317.82 USD
76304-066 76304-068 76304-070
Anti-IgG Rabbit Polyclonal Antibody (Biotin) PD-L1 Fc
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR