Anti-IgG Rabbit Polyclonal Antibody p16-INK-4a TAT

Supplier: Peprotech
500-P284T-50UG 500-P284T-100UG 500-P284T-1MG
76304-000EA 317.82 USD
76304-000 76304-002 76304-004
Anti-IgG Rabbit Polyclonal Antibody p16-INK-4a TAT
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR