Anti-IgG Rabbit Polyclonal Antibody TSLP

Supplier: Peprotech
500-P258-50UG 500-P258-100UG 500-P258-1MG
10777-334EA 339.8 USD
10777-334 10777-336 10777-338
Anti-IgG Rabbit Polyclonal Antibody TSLP
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR