Anti-IgG Rabbit Polyclonal Antibody Artemin

Supplier: Peprotech
500-P245-50UG 500-P245-100UG 500-P245-1MG
10777-194EA 339.8 USD
10777-194 10777-196 10777-198
Anti-IgG Rabbit Polyclonal Antibody Artemin
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR