EnGen Lba Cas12a (from Lachnospiraceae bacterium)
Supplier: New England Biolabs (NEB)
M0653S M0653T
76199-124EA 77 USD
76199-124 76199-126
EnGen Lba Cas12a (from Lachnospiraceae bacterium)
Enzymes
EnGen Lba Cas12a (Cpf1) from Lachnospiraceae bacterium ND20006 is a site-specific and programmable DNA endonuclease guided by a single 41-44 nucleotide guide RNA (gRNA).
  • T-rich TTTV PAM sequence opens up additional genomic regions for targeting
  • Shorter, 40-44 base guide RNA
  • Two nuclear localization signals for improved transport to the nucleus
  • 5' overhanging termini on cleavage products
  • Active from 16 to 48 °C
  • Heat inactivation 65 °C for 10 min
Targeting requires a gRNA complementary to the target site as well as a 5' TTTN protospacer adjacent motif (PAM) on the DNA strand opposite the target sequence. Cleavage by EnGen Lba Cas12a (Cpf1) occurs ~17 bases 3' of the PAM and leaves 5' overhanging ends.
Maintains activity at lower temperatures than the Acidaminococcus orthologs, permitting editing in ectothermic organisms such as zebra fish and xenopus.
EnGen Lba Cas12a has Simian virus 40 (SV40) T antigen nuclear localization sequences (NLS) at both the N and C-termini of the protein.
Información de suministro: Includes NEBuffer™ 2.1 (10X).