Sodium hydrogen carbonate 99.0-100.5% (dry basis), powder USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US


Management of Change (MOC) category = R

BS17-10 BS17-D1 BS17-D2 BAKRBS17-SA
JTBS17-10EA 126.92 USD
JTBS17-10 JTBS17-D1 JTBS17-D2 JTBS17SA
Sodium hydrogen carbonate 99.0-100.5% (dry basis), powder USP, Multi-Compendial, J.T.Baker®
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification Passes Test
USP - Limit of Sulfur Compounds ≤ 0.015 %
USP - Loss on Drying ≤ 0.25 %
USP - Insoluble Matter Passes Test
USP - Normal Carbonate Passes Test
USP - Limit of Ammonia ≤ 20 ppm
USP - Chloride (Cl) ≤ 0.015 %
USP - Arsenic (As) ≤ 2 ppm
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
EP/BP - Assay (NaHCO₃) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Carbonate (CO₃) Passes Test
EP/BP - Chloride (Cl) ≤ 150 ppm
EP/BP - Sulfate (SO₄) ≤ 150 ppm
EP/BP - Ammonium (NH₄) ≤ 20 ppm
EP/BP - Calcium (Ca) ≤ 100 ppm
EP/BP - Iron (Fe) ≤ 20 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR