Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US


Management of Change (MOC) category = R

BS14-10 BS14-D1 BS14-D2 BS14-SA BS14-ST BS14-D3
JTBS14-10EA 2122.24 USD
JTBS14-10 JTBS14-D1 JTBS14-D2 JTBS14-SA JTBS14-ST JTBS14-D3
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
MDL Number: MFCD00004679
CAS Number: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=81895d1047698a1dfe0d7ffb45301528644796fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets JPC (1997) Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₄H₁₁NO₃) 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification C Passes Test
USP - pH (1 in 20) 10.0 - 11.5
USP - Loss on Drying ≤ 1.0 %
USP - Residue on Ignition ≤ 0.1 %
USP - Melting Range(Lower) 168 - 172 °C
USP - Melting Range(Upper) 168 - 172 °C
EP/BP - Assay (C₄H₁₁NO₃) (dried basis) 99.0 - 100.5 %
EP/BP - Identification B (melting point) 168 - 174 °C
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 10.0 - 11.5
EP/BP - Related Substances ≤ 1.0 %
EP/BP - Chloride (Cl) ≤ 100 ppm
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying at 105°C ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JPC 1997 - Assay (C₄H₁₁NO₃) (dried basis) ≥ 99.0 %
JPC 1997 - Identification A Passes Test
JPC 1997 - Identification B Passes Test
JPC 1997 - pH 10.3 - 10.7
JPC 1997 - Melting Point 168 - 172 °C
JP - Clarity and Color of Solution Passes Test
JPC 1997 - Heavy Metals (as Pb) ≤ 8 ppm
JPC 1997 - Arsenic (As) ≤ 1.6 ppm
JPC 1997 - Loss on Drying ≤ 0.5 %
JPC 1997 - Residue on Ignition ≤ 0.10 %
Ultrapure Bioreagent Specifications
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR