Benzyl alcohol 98.0-100.5% NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

Management of Change (MOC) category = R

BS06-50 BS06-SA BS06-20
JTBS06-50CS 628.35 USD
JTBS06-50 JTBS06-SA JTBS06-20
Benzyl alcohol 98.0-100.5% NF, Multi-Compendial, J.T.Baker®
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Melting Pt: –15.4…–15.2 °C
Flash Pt: 100.4° C
MDL Number: MFCD00004599
CAS Number: 100-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (C₆H₅CH₂OH) 98.0 - 100.5 %
NF - Clarity of Solution Passes Test
NF - Color of Solution Passes Test
NF - Identification Passes Test
NF - Peroxide Value ≤ 5
NF - Benzaldehyde and Other Related Substances - Benzaldehyde ≤ 0.15 %
NF - Related Substances - Cyclohexylmethanol ≤ 0.10 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks ≤ 0.04 %
of Benzyl Alcohol ≤ 0.04 %
Greater Than RT of Benzyl Alcohol ≤ 0.3 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks ≤ 0.3 %
NF - Refractive Index, n²ºD 1.538 - 1.541
NF - Acidity Passes Test
NF - Residue on Evaporation ≤ 0.05 %
NF - Benzene ≤ 2 ppm
EP/BP - Assay (C₆H₅CH₂OH) 98.0 - 100.5 %
EP/BP - Identification Passes Test
EP/BP - Acidity Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Refractive Index, n²º^D 1.538 - 1.541
EP/BP - Benzaldehyde and Other Related Substances - Benzaldehyde ≤ 0.15 %
EP/BP - Related Substances - Cyclohexylmethanol ≤ 0.10 %
≤ 0.04 %
EP/BP - Benzaldehyde and Other Related Substances - Total Other Peaks ≤ 0.04 %
EP/BP - Benzaldehyde and Other Related Substances - TOTAL OTHER PEAKS ≤ 0.3 %
>RT OF BENZYL ALCOHOL ≤ 0.3 %
EP/BP - Peroxide Value ≤ 5
EP/BP - Residue on Evaporation ≤ 0.05 %
This product is not intended for use in preparing parenteral dosage
forms.
The Class 1 solvent Benzene is likely to be present below 2 ppm. The
actual analysis of Benzene is reported for each lot.
The Class 2 solvent Toluene is likely to be present and is process
controlled to <30 ppm.
No other Class 1, 2 or 3 residual solvents are likely to be present.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR