Labels 'Caution - Radioactive Materials', HCL Labels
Supplier: HCL Label
SGS-0053-0035 SGS-0053-0023 S32C-18-0011 S32C-18-0023 S32C-23-0023
103458-912PK 143 USD
103458-912 103458-970 103459-090 103459-092 103459-094
Labels 'Caution - Radioactive Materials', HCL Labels
Labels
Caution radioactive materials labels is durable and a great value.
  • Available in multiple sizes
Use this label to comply with regulations requiring laboratories to label small tools and equipment used to handle radioisotopes. Ideal to label pipettes, well plates, test tube racks, flasks, burets, clamps, and so on. These labels are laminated for extended durability that is to prevent fading from frequent handling and cleaning.