Clorox® Healthcare® Bleach Germicidal Cleaner
Supplier: Janitorial Supplies
CLO68978 CLO68970
21850-210CS 579.08 USD
21850-210 21850-204 21850-206
Clorox® Healthcare® Bleach Germicidal Cleaner
Cleaning Agents Disinfectants
Healthcare professionals trust Clorox® Healthcare® bleach germicidal cleaner to help them overcome infection control challenges in a constantly changing healthcare environment.
  • No pre-cleaning required
  • Designed for easy refilling to meet needs of heavy users
  • EPA registered to kill a total of 50 micro-organisms
  • Proven efficacy against outbreak-causing viruses Influenza A and B viruses, Measles virus and Norovirus
  • Kill HAI-causing pathogens including Clostridioides difficile spores, MRSA and VRE in 3 minutes or less
  • Broad Spectrum disinfection efficacy against over 50 pathogens
  • 3 minutes contact time to kill Clostridioides difficile spores - tested in the presence of 3-part organic soil load following the latest EPA testing recommendations (Nov. 2016)
  • Feature an anti-corrosion agent to ensure compatibility with healthcare surfaces
  • Tested for surface safety on common healthcare and office surfaces and equipment
Clorox® Healthcare® Bleach Germicidal Cleaner is a ready to use, 1 step cleaner- disinfectant containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings. This cleaner disinfectant is EPA registered to kill a total of 50 micro-organisms, including HAI causing bacteria like MRSA and VRE, outbreak causing viruses such as Influenza A and B viruses, Measles virus and Norovirus and fungi in 1 minute.
This disinfectant kill 99.9999% of Clostridioides difficile spores in 3 minutes, according to the current EPA recommended testing standard (June 2014). The ready-to-use disinfectant eliminates compliance risks and labor costs associated with dilatable disinfectants. Designed for easy refilling to meet needs of heavy users. This bleach solution features an anti-corrosion agent and is formulated for broad surface compatibility with healthcare surfaces including stainless steel, glass, plastic, porcelain, glazed ceramic, laminated surfaces and vinyl. Intended for use in healthcare settings. For use against bacteria, fungus, germs, grease, grime, mildew, mold, organic matter, virus, water-based soil. Works on ceramics, cement/concrete, enamel, fiberglass, glass, grout, laminates, metal, plastic, sealed/engineered stone, vinyl.
Certificaciones: Meets CDC and OSHA recommendations.