SelecTech Define Staining System, Leica
Supplier: Leica Biosystems
3801571 3801575 3801606 3801619 3803591 3802916 3801570 3802918 3803598 3803595
10015-076CS 230.92 USD
10015-076 10015-078 10015-080 10015-084 10015-086 10015-088 75810-352 75810-354 75810-358 75810-360 75839-344
SelecTech Define Staining System, Leica
Stains Histology Stains
SelecTech is a hematoxylin and eosin staining system consisting of Hematoxylin 560, Blue Buffer 8, Define, Alcoholic Eosin Y 515 and Eosin Trichrome 515. SelecTech Ready-To-Use Define aq is available in concentrates as well as ready-to-use formulas. Define is a refinement product designed to create crisp, nuclear detail and cosistency.
SelecTech standardizes the reagents and stains used in the hematoxylin and eosin staining procedure.
The built-in quality assurance parameters assure consistent results. The reagents and stains in SelecTech are designed to complement each other to produce a crisp, well delineated hematoxylin and eosin stain.